Learn More
Cayman Chemical Linoleic Acidbiotin
SDP
Supplier:  Cayman Chemical 100106231 MG
Encompass_Preferred
An affinity probe designed to allow linoleic acid to be detected in complexes with protein-binding partners such as fatty acid binding proteins
Catalog No. 50-288-8584