Learn More
Medchemexpress LLC α-Linolenic acid (Standard) | 463-40-1 | 278.43 | 50 MG

Supplier: Medchemexpress LLC HYN0728R50MG
α-Linolenic acid (Standard) is the analytical standard of α-Linolenic acid. This product is intended for research and analytical applications. It is an essential fatty acid isolated from Perilla frutescens that cannot be synthesized by humans. It can affect the process of thrombotic through the modulation of PI3K/Akt signaling, possesses anti-arrhythmic properties, and is related to cardiovascular disease and cancer.
- Analytical standard for α-Linolenic acid.
- Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
- Isolated from Perilla frutescens.
- Essential fatty acid.
- Affects thrombotic processes via PI3K/Akt signaling.
- Has anti-arrhythmic properties.
- Related to cardiovascular disease and cancer.
- Appearance: Liquid (density: 0.887 g/cm3), colorless to light yellow.
- Purity: 99.32%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.