Learn More
Cayman Chemical LInoleoyl GlycIn 10mg
SDP
Supplier:  Cayman Chemical 900032610 MG
Encompass_Preferred
An endogenous homolog of arachidonyl glycine that inhibits AEA hydrolysis in FAAH-containing N18TG2 cell membranes with an IC50 value of 25 µM
Catalog No. 50-289-7012