Learn More
Cayman Chemical LInoleoylLcarnItInchLRI 10mg
SDP
Supplier:  Cayman Chemical 2656010 MG
Encompass_Preferred
A long-chain acylcarnitine; levels are increased following high-dose (200 mg/kg) administration of isoniazid in mice
Catalog No. 50-293-5246