Learn More
Medchemexpress LLC Linoleyl alcohol | 506-43-4 | 266.46 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0056271G
Encompass_Preferred
Linoleyl alcohol is a fatty alcohol, a structural analog of Linoleic acid that lacks an α-carboxyl group. It is intended for research use only and has been shown to be effective in weight-loss promotion and metabolic syndrome management in animal studies.
  • Fatty alcohol, structural analog of linoleic acid
  • Lacks an α-carboxyl group
  • Used in research applications
  • Dioxygenation by potato tuber lipoxygenase leads to the formation of hydroperoxyoctadecadien-1-ols
  • Esters with Gallic acid and (-)-Epigallocatechin promote weight loss and metabolic syndrome management in animal models
Catalog No. 50-004-49317