Learn More
Medchemexpress LLC Linoleyl alcohol | 506-43-4 | 266.46 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00562710G
Encompass_Preferred
Linoleyl alcohol is a fatty alcohol, a structural analog of Linoleic acid that lacks an a-carboxyl group. This product is a colorless to light yellow liquid.
  • Pure form stable for 3 years at -20°C
  • In solvent stable for 6 months at -80°C and 1 month at -20°C
  • Harmful if swallowed
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Soluble in DMSO (100 mg/mL)
  • Used in research on dioxygenation by potato tuber lipoxygenase
  • Esters of Linoleyl alcohol may promote weight-loss and manage metabolic syndrome
Catalog No. 50-004-45371