Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linoleyl alcohol | 506-43-4 | MFCD00056667 | 266.46 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0056275G

Encompass_Preferred

Linoleyl alcohol is a fatty alcohol and a structural analog of Linoleic acid, lacking an α-carboxyl group. This product is intended for research use only and is not sold to patients.

  • Provided as a colorless to light yellow liquid
  • Molecular weight of 266.46
  • Purity of 99.8%
  • Soluble in DMSO (100 mg/mL)
  • Supports research on weight-loss and metabolic syndrome management
  • Store pure form at -20°C for up to 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-47321


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.