Learn More
Medchemexpress LLC Linsidomine hydrochloride | 16142-27-1 | MFCD08705735 | 100.0% | 206.63 g/mol | C6H11ClN4O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10120010MM 1ML
Encompass_Preferred
Linsidomine hydrochloride is a research-grade peroxynitrite donor that spontaneously generates reactive oxygen and nitrogen species for laboratory studies. It is used to model oxidative and nitrosative stress, investigate vascular smooth muscle and platelet biology, and can be supplied as a ready-to-use solution for immediate experimental use.
  • Spontaneous generator of reactive oxygen and nitrogen species.
  • Acts as a peroxynitrite donor for oxidative stress studies.
  • Useful in vascular biology and platelet aggregation research.
  • Available as a 10 mM solution in DMSO, 1 mL, for ready-to-use experiments.
  • Also supplied as solids in multiple mass quantities for formulation flexibility.
  • High analytical purity suitable for research applications.
Catalog No. 50-002-11640