Learn More
Medchemexpress LLC HY-10191 10mg Medchemexpress, Linsitinib CAS:867160-71-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1019110MG
Encompass_Preferred
Medchemexpress, HY-10191 10mg Linsitinib CAS:867160-71-2 Linsitinib (OSI-906) is a potent, selective and orally bioavailable dual inhibitor of the IGF-1 receptor and insulin receptor (IR) with IC50s of 35 and 75 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8687