Learn More
TARGETMOL CHEMICALS INC LINSITINIB 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T601725MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OSI-906 (Linsitinib (OSI-906)) is an orally bioavailable small molecule inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) with potential antineoplastic activity. Linsitinib selectively inhibits IGF-1R which may result in the inhibition of tumor cell proliferation and the induction of tumor cell apoptosis. Overexpressed in a variety of human cancers IGF-1R stimulates cell proliferation enables oncogenic transformation and suppresses apoptosis. purity: 99%
Catalog No. 50-257-6627