Learn More
Selleck Chemical LLC Linsitinib (OSI-906) 10mM/1mL 867160-71-2
SDP
Supplier:  Selleck Chemical LLC S109110MM/1ML
Encompass_Preferred
Linsitinib (OSI-906) is a selective inhibitor of IGF-1R with IC50 of 35 nM in cell-free assays; modestly potent to InsR with IC50 of 75 nM, and no activity towards Abl, ALK, BTK, EGFR, FGFR1/2, PKA etc. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0907