Learn More
Medchemexpress LLC Linuron-d6 | 1219804-76-8 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1866S5MG
Encompass_Preferred
Linuron-d6 is the deuterium labeled Linuron, a phenylurea herbicide widely used to control the growth of grass and weeds in various agricultural crops and orchards. It functions as a photosystem II inhibitor and a competitive androgen receptor (AR) antagonist with a Ki of 100 μM. Linuron has been observed to exhibit reproductive toxicity in animals, acting as an endocrine disruptor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during the drug development process and have the potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-12143