Learn More
Cayman Chemical LInzolId-d3 500ug
SDP
Supplier:  Cayman Chemical 25038500 UG
Encompass_Preferred
An internal standard for the quantification of linezolid by GC- or LC-MS
Catalog No. 50-293-4494