Learn More
Medchemexpress LLC HY-A0070A 100mg , Liothyronine CAS:6893-02-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0070A100MG
Encompass_Preferred
HY-A0070A 100mg Liothyronine CAS: 6893-02-3 Liothyronine is an active form of thyroid hormone. Liothyronine is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0275