Learn More
Apexbio Technology LLC Liothyronine Sodium 55-06-1 500mg
SDP
Supplier:  Apexbio Technology LLC B1779500
Encompass_Preferred
Liothyronine Sodium (55-06-1) is a small-molecule agonist targeting nuclear thyroid hormone receptors hTR and hTR It is designed to activate these receptors thereby modulating transcriptional regulation of genes involved in energy metabolism protein synthesis lipid metabolism and thermogenesis Liothyronine Sodium exerts its biological activity primarily through binding and activating hTR and hTR In in vitro studies Liothyronine Sodium demonstrates agonist activity with EC50 values of 2 33 nM for both hTR and hTR Based on these pharmacological properties Liothyronine Sodium holds research potential in investigating thyroid-related metabolic pathways receptor-ligand interactions and molecular mechanisms underlying thyroid hormone regulation and metabolic disorders
Catalog No. 50-000-94317