Learn More
Medchemexpress LLC Liothyronine (Standard) | 6893-02-3 | 98.6% | C15H12I3NO4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA0070AR50MG
Encompass_Preferred
Liothyronine (Standard) is the analytical standard of Liothyronine, an active form of thyroid hormone. It is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.
  • Intended for research and analytical applications.
  • Used as a reference standard in assays.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-004-32770