Learn More
Medchemexpress LLC Lipid 14 | 2430034-05-0 | C56H110N2O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15337710MM/1ML
Encompass_Preferred
Lipid 14 is an ionizable amino lipid that facilitates the delivery of mRNA into target cells. It significantly enhances the in vivo expression efficiency and immunogenicity of mRNA. This lipid can be utilized for the synthesis of lipid nanoparticles (LNPs) and has applications in studies related to SARS-CoV-2 and Yersinia pestis.
  • Delivers mRNA into target cells.
  • Significantly enhances in vivo expression efficiency of mRNA.
  • Significantly enhances the immunogenicity of mRNA.
  • Can be used to synthesize lipid nanoparticles (LNPs).
  • Applicable for studies of SARS-CoV-2 and Yersinia pestis.
Catalog No. 50-003-72745