Learn More
Echelon Biosciences Research Labs Lipid 319, 100 mg, CAS Number: 1351586-50-9
SDP
Supplier:  Echelon Biosciences Research Labs N1319100MG
Encompass
Lipid 319 is an ionizable and biodegradable lipid. Lipid 319 nanoparticles have better delivery efficacy and biodegradability than DLin-MC3-DMA in the delivery of mRNA in vivo. MC3 is the key delivery nanoparticle component of the first siRNA drug Onpattro.
Catalog No. NC2753042