Learn More
Cayman Chemical LIpId A Monphosphoryl from1mg
SDP
Supplier:  Cayman Chemical 258481 MG
Encompass_Preferred
A TRIF-biased TLR4 agonist; induces T cell clonal expansion and TRIF-dependent production of G-CSF, IL-10, IP-10, and MCP-1 but has no effect on MyD88-dependent production of the inflammatory cytokines IL-1β, IL-6, MIP-1α, and IFN-γ in mice
Catalog No. 50-293-4330