Learn More
Medchemexpress LLC Lipocalin-2/NGAL Cy 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP791145UG
Encompass_Preferred
Lipocalin-2/NGAL is a multifaceted iron transporter involved in multiple biological processes including apoptosis innate immunity and kidney development Lipocalin-2 dynamically affects cellular iron concentration through binding to the siderophore 2 3-dihydroxybenzoate Lipocalin-2/NGAL Protein Cynomolgus (CHO His) is the recombinant cynomolgus-derived Lipocalin-2/NGAL protein expressed by CHO with C-His labeled tag
Catalog No. 50-001-94579