Learn More
Medchemexpress LLC Lipocalin-2/NGAL Mo 10ug
SDP
Supplier:  Medchemexpress LLC HYP70658A10UG
Encompass_Preferred
Recombinant mouse Lipocalin-2 (NGAL) expressed in HEK293 cells and supplied as a filtered, stabilized solution with a C-terminal 6xHis tag. The preparation is formulated for stability and lot-to-lot consistency, and is intended for research use in biochemical and immunological assays, antigen preparation, and in vitro functional studies.
  • Supplied as a 0.22 μm filtered solution in a stabilizing buffer.
  • Expressed in mammalian cells for relevant post-translational modifications.
  • Contains a C-terminal 6xHis tag for detection and purification.
  • Available in small, research-friendly sizes suitable for assay development.
  • Suitable for biochemical assays, immunoassays, and antigen preparation.
  • Provided with batch-specific certificate of analysis and safety documentation.
Catalog No. 50-001-84203