Learn More
Medchemexpress LLC Lipocalin-2/NGAL Mo 500ug
SDP
Supplier:  Medchemexpress LLC HYP70658A500UG
Encompass_Preferred
Recombinant mouse Lipocalin-2 (NGAL) is a glycosylated protein expressed in HEK293 cells, purified and supplied lyophilized with a C-terminal 6xHis tag. It is intended for research use in biochemical assays, antibody validation, and functional studies, and is supplied with confirmed purity and recommended frozen storage for long-term stability.
  • Glycosylated protein expressed in mammalian cells for native-like post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and sensitive detection.
  • High purity (>95%) as verified by reducing SDS-PAGE.
  • Available in multiple sizes, including a 500 μg format.
  • Lyophilized formulation stable for frozen storage; aliquot and freeze for extended use.
Catalog No. 50-001-85328