Learn More
Medchemexpress LLC HY-116788 10mg Medchemexpress, Lipofermata CAS:297180-15-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11678810MG
Encompass_Preferred
Medchemexpress, HY-116788 10mg Lipofermata CAS:297180-15-5 Lipofermata is a fatty acid transport proteins (FATP) inhibitor that abrogates lipid transport into melanoma cells and reduces melanoma growth and invasion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9439