Learn More
Selleck Chemical LLC Lipofermata-E1533-5MG
SDP
Supplier:  Selleck Chemical LLC E15335MG
Encompass_Preferred
Lipofermata is an inhibitor of fatty acid transport protein 2 (FATP2) that demonstrates fatty acid transport inhibition with an IC50 of 4 84 M in Caco-2 cells It may offer therapeutic potential for diseases where lipotoxicity is a key factor
Catalog No. 50-313-8719