Learn More
Selleck Chemical LLC Lipopolysaccharides-1mg
SDP
Supplier:  Selleck Chemical LLC S78501MG
Encompass_Preferred
Lipopolysaccharides (LPSs, Endotoxin,LPS) is an endotoxin derived from the outer leaflet of the outer membrane of Gram-negative bacteria. Lipopolysaccharides consists of an antigen O-specific chain, a core oligosaccharide and lipid A. Lipopolysaccharides is a pathogenic associated molecular pattern (PAMP) that activates the immune system. Lipopolysaccharides activates TLR-4 on immune cells. Lipopolysaccharides induces secretion of cell migrasome.
Catalog No. 50-238-9722