Learn More
Cusabio Technology LLC Human Lipopolysaccharides,LPS ELISA Kit
Supplier:  Cusabio Technology LLC CSBE09945H
Encompass_Preferred
This Human LPS ELISA Kit was designed for the quantitative measurement of Human LPS protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 6.25 pg/mL-400 pg/mL and the sensitivity is 1.56 pg/mL. One kit contains 96 wells.
Catalog No. NC2952932