Learn More
Medchemexpress LLC Lipopolysaccharides, from P. aeruginosa 10 | 10 MG

Supplier: Medchemexpress LLC HYD1056E10MG
Lipopolysaccharides from Pseudomonas aeruginosa 10 are endotoxin preparations used to activate Toll-like receptor 4 (TLR4) in in vitro immune assays and endotoxin challenge studies. The material is supplied as a white to off-white solid with high endotoxin potency and low protein contamination, suitable as a positive control for immune stimulation.
- Source: Pseudomonas aeruginosa strain 10.
- High endotoxin potency (600,000 EU/mg) for consistent immune activation.
- Low protein contamination (0.82% protein content by COA).
- White to off-white solid form for easy handling and storage.
- Storage recommendations: 4°C sealed; in solvent store -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.