Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides P100MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L9143100MG
Encompass
This product is phenol extracted from Pseudomonas aeruginosa serotype 10.22 The source strain is ATCC 27316.
Catalog No. 50-290-4014