Learn More
Cusabio Technology LLC Mouse Lipopolysaccharides(LPS) ELISA Kit
Supplier:  Cusabio Technology LLC CSBE13066M
Encompass_Preferred
This Mouse LPS ELISA Kit was designed for the quantitative measurement of Mouse LPS protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 0.156 ng/mL-10 ng/mL and the sensitivity is 0.039 ng/mL . One kit contains 96 wells.
Catalog No. NC2952951