Learn More
Cayman Chemical Lipoxin A5 Fatty Acids and Der
SDP
Supplier:  Cayman Chemical 1001145350 UG
Encompass_Preferred
A 5-series LO derived from EPA; contracts pulmonary parenchymal strips from guinea pig with similar potency to that of LXA4 and LXB4, yet does not exert the vasodilatory effects on aortic smooth muscle exhibited by LXA4 and LXB4
Catalog No. 50-289-0999