Learn More
Medchemexpress LLC HY-12726 10mg , Liproxstatin-1 CAS:950455-15-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1272610MG
Encompass_Preferred
HY-12726 10mg Liproxstatin-1 CAS: 950455-15-9 Liproxstatin-1 is a potent ferroptosis inhibitor and inhibits ferroptotic cell death (IC50=22 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9134