Learn More
Chemglass Life Sciences Extractor, Liquid-Liquid, Continous, 45/50, 24/40
SDP
Supplier:  Chemglass Life Sciences CG136001
Encompass_Preferred
Extractor, Liquid-Liquid, Continuous, 45/50, 24/40. Extractor designed for EPA priority pollutant samples where the extracting solvent is heavier than water. For samples of 1 liter plus approximately 200mL of extracting solvent. Extractor has a 45/50 top outer joint and a 24/40 inner joint on the uptake tube. Flask and condenser must be ordered separately.
Catalog No. CG136001