Learn More
Electron Microscopy Sciences Paper Liquid Wicks, 28 mm, Size Extra-Fine
SDP
Supplier:  Electron Microscopy Sciences 60160XF
Encompass
Theses paper wicks offer a great deal of absorbency with small tips that foam-tipped or cotton-tipped applicators can not substitute. Each box contains 300, 28 mm long high quality, tapered paper wicks. Order Extra Fine (XF) for removing liquid formed around your crystal.
Catalog No. 50-905-3391