Learn More
Medchemexpress LLC Liquiritigenin | 578-86-9 | 256.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN037750MG
Encompass_Preferred
Liquiritigenin is a flavanone isolated from *Glycyrrhiza uralensis*. It functions as a highly selective estrogen receptor β (ERβ) agonist, with an EC50 of 36.5 nM for activating ERE tk-Luc. This compound is intended for research use only.
  • Acts as a selective estrogen receptor β (ERβ) agonist (EC50 of 36.5 nM).
  • Activates ERE tk-Luc in U2OS cells transfected with ERβ.
  • Recruits steroid receptor coactivator-2 (SRC-2) to target genes in ERβ cells.
  • Prevents methylglyoxal-induced cell death in MC3T3-E1 cells.
  • Reduces pro-inflammatory cytokines in serum and hippocampus in vivo.
  • Does not stimulate uterine size or tumorigenesis of MCF-7 breast cancer cells in vivo.
Catalog No. 50-003-90059