Learn More
Medchemexpress LLC Liquiritigenin-7-O-β-D-glucopyranosyl-(1→2)-β-D-apiofuranoside | 135432-48-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN69865MG
Encompass_Preferred
Liquiritigenin-7-O-β-D-glucopyranosyl-(1→2)-β-D-apiofuranoside, also known as Liquiritigenin-7-apiosylglucoside, is a flavonoid that has been isolated from the roots of Glycyrrhiza. It exhibits weaker cytotoxicity against several tumor cells and normal cells.
  • Isolated from the roots of Glycyrrhiza.
  • Exhibits weaker cytotoxicity against several tumor cells and normal cells.
Catalog No. 50-004-18524