Learn More
Med Vet International Liquivet Tissue Adhesive, 8 Seconds, Vet Use, Bottle, 3g, 6 Applicators
Supplier:  Med Vet International VVG3SC
Encompass_Preferred
Veterinary use only. LiquiVet monomeric formulation that forms a thin, waterproof adhesion to bond tissue in a variety of vet procedures. Comes in 3gr dispensing bottles with 6 applicators. Fast-acting Cyanoacrylate based tissue adhesive for wound care. Polymerizes in 8 seconds. Ideal for skin closure and to seal nerve endings and capillary beds after cat declaw procedures. Excellent for quick repair of minor lacerations, abrasions, and other minor wounds. Superior performance in the time necessary to close a wound. For external lacerations, not to be used internally. May be alternative to products like ZipStitch instant stitches and Strips For skin closure Best suited for small, superficial lacerations Quick and virtually painless to use Wears off in the days and weeks Reduce the chances of an infection
Catalog No. 50-252-2143