Learn More
Medchemexpress LLC HY-P0014 10mg , Liraglutide CAS:204656-20-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYP001410MG
Encompass_Preferred
Medchemexpress, HY-P0014 10mg Liraglutide CAS:204656-20-2 Formula:C172H265N43O51 GLP-1 receptor Purity:98% Liraglutide is a glucagon-like peptide-1 ( GLP-1 ) receptor agonist used clinically to treat type 2 diabetes mellitus. Sequence: His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-{N6-[N-(1-oxohexadecyl)-L-γ-glutamyl]-Glu}-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Arg-Gly-Arg-Gly. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4050