Learn More
Medchemexpress LLC Liraglutide-13C6,15N TFA | 204656-20-2 | >97.1% | 3758.15 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP0014S21MG
Encompass_Preferred
Liraglutide-13C6,15N TFA is a 13C and 15N labeled version of Liraglutide. Liraglutide is known as a glucagon-like peptide-1 (GLP-1) receptor agonist, primarily studied for its role in type 2 diabetes mellitus.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-004-26523