Learn More
Medchemexpress LLC Liraglutide (TFA) | 204656-20-2 | 99.6% | 3751.20 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0014B10MG
Encompass_Preferred
Liraglutide (TFA) is a glucagon-like peptide 1 receptor (GLP-1) agonist. It activates GLP-1, leading to insulin release in the presence of increased glucose concentration and reduces glucagon secretion in a glucose-dependent manner. It can be used in research on type 2 diabetes.
  • Agonist of glucagon-like peptide 1 receptor (GLP-1)
  • Activates GLP-1
  • Leads to insulin release in the presence of increased glucose concentration
  • Reduces glucagon secretion in a glucose-dependent manner
  • Used in research on type 2 diabetes
Catalog No. 50-004-26757