Learn More
Medchemexpress LLC Lisaftoclax | 2180923-05-9 | 99.8% | 100 MG

Supplier: Medchemexpress LLC HY129179100MG
Lisaftoclax (compound 6) is a dual Bcl-2 and Bcl-xl inhibitor with anti-tumor activity. It exhibits IC50 values of 2 nM for Bcl-2 and 5.9 nM for Bcl-xl. In vitro, Lisaftoclax shows IC50 values of 5.5 nM and 6.4 nM in Bcl-2 dependent RS4;11 cells and Bcl-xl dependent Molm13 cells.
- Dual Bcl-2 and Bcl-xl inhibitor.
- Demonstrates anti-tumor activity.
- High purity of 99.8%.
- Light yellow to yellow solid appearance.
- Consistent with structure by 1H NMR and MS.
- Stable as powder for 3 years at -20°C, 2 years at 4°C.
- Stable in solvent for 6 months at -80°C, 1 month at -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.