Learn More
TARGETMOL CHEMICALS INC LISAFTOCLAX 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T104835MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lisaftoclax (Bcl-2/Bcl-xl inhibitor 1) is an oral dual inhibitor of Bcl-2 and Bcl-xl with IC50 values of 2 nM and 5.9 nM for Bcl-2 and Bcl-xl respectively. Lisaftoclax is used to treat chronic lymphocytic leukemia (CLL) by inhibiting the action of the BCL-2 protein which promotes the death of leukemia cells. purity: 98%
Catalog No. 50-258-1718