Learn More
Medchemexpress LLC Lisinopril (dihydrate) | 83915-83-7 | 100.0% | 441.52 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY18206500MG
Encompass_Preferred
Lisinopril dihydrate (MK-521 dihydrate) is an angiotensin-converting enzyme inhibitor, used in the treatment of hypertension, congestive heart failure, and heart attacks.
  • Targets angiotensin-converting enzyme (ACE).
  • Used in cardiovascular disease research.
  • Used in cancer research.
Catalog No. 50-003-87731