Learn More
Enzo Life Sciences (±)-Lisofylline (10mg). CAS: 6493-06-7
SDP
Supplier:  Enzo Life Sciences BMLLP1020010
Encompass_Preferred
A potent inhibitor of phosphatidic acid generation (IC50=0.6 µM). It protects mice from endotoxic shock and attenuates sepsis-induced acute lung injury in the pig. Anti-inflammatory agent. Enhances glucose-stimulated insulin secretion. Inhibits IL-12-mediated STAT4 activation. Purity: ≥98% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-1153