Learn More
TARGETMOL CHEMICALS INC Research Chemical 25mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T2172925MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (±)-Lisofylline is an enantiomer-specific, alkyl-substituted methylxanthine, which has specific and potent activity in down-regulating leukocyte activation. (±)-Lisofylline is an anti-inflammatory agent. purity: 99%
Catalog No. 50-303-8143