Learn More
Medchemexpress LLC LIT-001 | 2245072-21-1 | 98.6% | 645.70 | C30H34F3N7O4S | 50MG
SDP
Supplier:  Medchemexpress LLC HY124733A50MG
Encompass_Preferred
LIT-001 is a nonpeptide oxytocin receptor (OT-R) agonist used as a research tool compound. It exhibits OT-R activity (EC50 = 55 nM; Ki = 226 nM) and has been reported to improve social interaction in a mouse model. The compound is a white to off-white solid (molecular formula C30H34F3N7O4S, MW 645.70), provided with analytical documentation, and is typically stored at -20 °C. It is highly soluble in DMSO for in vitro use.
  • Nonpeptide oxytocin receptor agonist with EC50 = 55 nM and Ki = 226 nM.
  • Demonstrated improvement of social interaction in a mouse model.
  • High purity suitable for in vitro studies.
  • High solubility in DMSO enables straightforward formulation.
  • Provided with analytical documentation such as COA, LC-MS, and H NMR.
  • Solid form stable when stored at -20 °C under sealed, dry conditions.
Catalog No. 50-002-37174