Learn More
Selleck Chemical LLC LIT-927 S8813-5MG
SDP
Supplier:  Selleck Chemical LLC S88135MG
Encompass_Preferred
LIT-927 is a novel neutraligand of CXCL12 with Ki value of 267 nM for inhibition of Texas red-labeled CXCL12 (CXCL12-TR) binding It shows high selectivity toward CXCL12 vs other chemokines also involved in asthma
Catalog No. 50-217-3393