Learn More
Cayman Chemical LItchI chIns AtrnorIn 10mg
SDP
Supplier:  Cayman Chemical 2810610 MG
Encompass_Preferred
A depside lichen metabolite; active against the bacteria B. cereus, B. subtilis, S. aureus, S. faecalis, P. vulgaris, L. monocytogenes, and A. hydrophila (MICs = 1.67, 0.38, 26.7, 13.4, 3.34, 9.83, and 1.67 mM, respectively), the fungi C. albicans and C. glabrata (MIC = 26.7 mM for both), as well as the mycobacterium M. aurum (MIC = 250 UG/ml); cytotoxic to A270, HL-60, and Jurkat cancer cells (IC50s = 197.9, 93.5, and 181.6 µM, respectively) but not HeLa, MCF-7, SK-BR-3, or HT-29 cancer cells (IC50s = >200 µM); inhibits acetic acid-induced writhing in mice at 200 and 400 mg/kg; reduces paw licking and biting in the second, but not first, phase of the formalin test when administered 30 minutes prior to formalin in mice at 200 and 400 mg/kg
Catalog No. 50-294-1268