Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Ship-Lite Flat Envelope, White, 10 x 13 in, 100/Case

Supplier:  MSC 39577234

Encompass

This flat envelope is designed for shipping and packaging applications, ensuring mailed materials are transported safely.

  • White color
  • Measures 10 x 13 inch
  • Made from 26 lb reinforced paper
  • Packaged as 100 per case

Catalog No. 50-191-8137


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.