Learn More
Medchemexpress LLC Cholan-24-oic acid, 3-[(2S)-2-amino-3-carboxy-1-oxopropoxy]-, (3α,5β)- | 881179-02-8 | 98.0% | 491.66 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112415
Encompass_Preferred
Lith-O-Asp is a sialyltransferase (ST) inhibitor, with IC50 values ranging from 12-37 μM against ST3Gal I, ST3Gal III, and ST6GalI. It inhibits α-2,3- and α-2,6-sialyltransferases, decreasing sialic acid transfer to targeted glycoproteins. Research shows potential in suppressing cancer angiogenesis and metastasis, with treated mice exhibiting fewer lung metastases in a 4T1-Luc breast cancer model.
  • Sialyltransferase (ST) inhibitor
  • IC50 values of 12-37 μM against ST3Gal I, ST3Gal III, and ST6GalI
  • No apparent growth inhibition effect on different cancer cell lines at tested doses (10, 30, and 60 μM)
  • Decreases expression of cell surface α-2,3- and α-2,6-sialylated antigens
  • Suppresses FAK/paxillin signaling and cancer angiogenesis and metastasis pathways in vivo
Catalog No. 50-004-40076