Learn More
Strem, An Ascensus Company CAS# 16853-85-3. 50g. Lithium aluminum hydride, powder, 95%. MFCD00011075
SDP
Supplier:  Strem, An Ascensus Company 93039350G
Encompass
CAS# 16853-85-3. 50g. Lithium aluminum hydride, powder, 95%. MFCD00011075. Molecular Weight: 37.94. Molecular Formula: LiAlH4. Color/form: gray pwdr. Strem# 93-0393. http://www.strem.com/catalog/v/93-0393/
Catalog No. 50-901-13693